Thermo Scientific Chemicals

Methyl stearate, mixture of homologs, Thermo Scientific Chemicals

Catalog number: 304220025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Methyl stearate, mixture of homologs, Thermo Scientific Chemicals

Catalog number: 304220025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 304220025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-61-8
IUPAC Name
methyl octadecanoate
Molecular Formula
C19H38O2
InChI Key
HPEUJPJOZXNMSJ-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC(=O)OC
Appearance (Form)
Powder or crystals or fused solid
Acid value
=<1 mg KOH/g
GC
>=90 % (methyl stearate)
Saponification value
186 to 208 mg KOH/g
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text