Thermo Scientific Chemicals

Cycloheptene, 95%, Thermo Scientific Chemicals

Catalog number: 304260250
25 g, Each
Thermo Scientific Chemicals

Cycloheptene, 95%, Thermo Scientific Chemicals

Catalog number: 304260250
25 g, Each
Quantity
Catalog number: 304260250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
628-92-2
IUPAC Name
cycloheptene
Molecular Formula
C7H12
InChI Key
ZXIJMRYMVAMXQP-UHFFFAOYSA-N
SMILES
C1CCC=CCC1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=94 %
Stabilizer
0.1 % BHT

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text