1x1 image pixel for data collection
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Thermo Scientific Chemicals

Palmitoyl chloride, 98%

CAS: 112-67-4 | C16H31ClO | 274.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 304291000
Price (USD)
62.65
Special offer
Online exclusive
Ends: 26-Sep-2025
77.20
Save 14.55 (19%)
Each
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 26-Sep-2025
77.20
Save 14.55 (19%)
Each
Add to cart
Chemical Identifiers
CAS112-67-4
IUPAC Namehexadecanoyl chloride
Molecular FormulaC16H31ClO
InChI KeyARBOVOVUTSQWSS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Acidity=<1 % (as palmitic acid)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Titration Argentometric98 to 101 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link