Thermo Scientific Chemicals

Palmitoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 304295000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Palmitoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 304295000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 304295000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-67-4
IUPAC Name
hexadecanoyl chloride
Molecular Formula
C16H31ClO
InChI Key
ARBOVOVUTSQWSS-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(Cl)=O
Titration Argentometric
98 to 101 %
Refractive index
1.4510 to 1.4530 (20°C, 589 nm)
GC
>=97.5 %
Acidity
=<1 % (as palmitic acid)
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text