Thermo Scientific Chemicals

2-Methylbutyraldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 304472500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Methylbutyraldehyde, 95%, Thermo Scientific Chemicals

Catalog number: 304472500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 304472500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-17-3
IUPAC Name
2-methylbutanal
Molecular Formula
C5H10O
InChI Key
BYGQBDHUGHBGMD-UHFFFAOYNA-N
SMILES
CCC(C)C=O
Infrared spectrum
Conforms
GC
>=94 %
Refractive index
1.3890 to 1.3950 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text