Thermo Scientific Chemicals

Bromophenol Red, pure

Catalog number: 304550050
5 g, Each
Thermo Scientific Chemicals

Bromophenol Red, pure

Catalog number: 304550050
5 g, Each
Quantity
Catalog number: 304550050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2800-80-8
IUPAC Name
3,3-bis(3-bromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C19H12Br2O5S
InChI Key
OYCLSQDXZMROJK-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Br)=C1
Solubility
(1 % in ethanol) Clear
Appearance (Form)
Powder
Loss on drying
=<5 % (110°C)
Visual transition interval
from pH 5.2 (yellow-red) to pH 7.0 (purple)
Appearance (Color)
Red-brown to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text