Bromophenol Red, pure
Bromophenol Red, pure
Thermo Scientific Chemicals

Bromophenol Red, pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 304550250
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
Add to cart
Bromophenol Red, pure
Catalog number304550250
Price (USD)/ Each
249.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
294.00 
Save 44.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2800-80-8
IUPAC Name3,3-bis(3-bromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H12Br2O5S
InChI KeyOYCLSQDXZMROJK-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(1 % in ethanol) Clear
Appearance (Form)Powder
Loss on drying=<5 % (110°C)
Visual transition intervalfrom pH 5.2 (yellow-red) to pH 7.0 (purple)
Appearance (Color)Red-brown to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only