1x1 image pixel for data collection
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure

Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg | CAS: 18154-32-0 | C10H12FeN2NaO8·3H2O | 421.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 304680010
Price (USD)
438.65
Online Exclusive
487.00
Save 48.35 (10%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
438.65
Online Exclusive
487.00
Save 48.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS18154-32-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=97.5 %
Appearance (Color)Yellow-brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Share catalog number, name or link