Thermo Scientific Chemicals

Methyl 2-chloro-4-fluorophenylacetate, 98%, Thermo Scientific Chemicals

Catalog number: 304780050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Methyl 2-chloro-4-fluorophenylacetate, 98%, Thermo Scientific Chemicals

Catalog number: 304780050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 304780050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
214262-88-1
IUPAC Name
methyl 2-(2-chloro-4-fluorophenyl)acetate
Molecular Formula
C9H8ClFO2
InChI Key
DFTYYBRLWCAUHP-UHFFFAOYSA-N
SMILES
COC(=O)CC1=CC=C(F)C=C1Cl
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.5060 to 1.5080 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text