4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 305000050
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
Add to cart
4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals
Catalog number305000050
Price (USD)/ Each
26.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.00 
Save 4.35 (14%)
-
Add to cart
Chemical Identifiers
CAS1120-97-4
IUPAC Name4-methyl-1,3-dioxane
Molecular FormulaC5H10O2
InChI KeyINCCMBMMWVKEGJ-UHFFFAOYNA-N
SMILESCC1CCOCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4140 to 1.4160 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only