Thermo Scientific Chemicals

4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals

Catalog number: 305000500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals

Catalog number: 305000500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305000500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1120-97-4
IUPAC Name
4-methyl-1,3-dioxane
Molecular Formula
C5H10O2
InChI Key
INCCMBMMWVKEGJ-UHFFFAOYNA-N
SMILES
CC1CCOCO1
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4140 to 1.4160 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text