Thymoquinone, 99%, Thermo Scientific™
Thymoquinone, 99%, Thermo Scientific™
Thermo Scientific™

Thymoquinone, 99%, Thermo Scientific™

 
Have Questions?
Catalog number 305070010
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
Add to cart
Thymoquinone, 99%, Thermo Scientific™
Catalog number305070010
Price (USD)/ Each
43.65
Online exclusive
47.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
CAS490-91-5
IUPAC Name2-methyl-5-(propan-2-yl)cyclohexa-2,5-diene-1,4-dione
Molecular FormulaC10H12O2
InChI KeyKEQHJBNSCLWCAE-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)Yellow to orange to brown
Melting point44°C to 47°C
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only