N-Methylphthalimide, 98%, Thermo Scientific Chemicals
N-Methylphthalimide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methylphthalimide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 305100250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS550-44-7
IUPAC Name2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC9H7NO2
InChI KeyZXLYYQUMYFHCLQ-UHFFFAOYSA-N
SMILESCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder or crystals
Melting point132°C to 136°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only