Thermo Scientific Chemicals

(2-Iodoethyl)benzene, 97%, stabilized with copper, Thermo Scientific Chemicals

Catalog number: 305250050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

(2-Iodoethyl)benzene, 97%, stabilized with copper, Thermo Scientific Chemicals

Catalog number: 305250050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 305250050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17376-04-4
IUPAC Name
(2-iodoethyl)benzene
Molecular Formula
C8H9I
InChI Key
KVTHPKXDLVYNCH-UHFFFAOYSA-N
SMILES
ICCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to red to brown
Infrared spectrum
Conforms
GC
>=96.0 %
Stabilizer
Copper
Refractive index
1.5990 to 1.6030 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text