Appearance (Color)Clear colorless to yellow to red to brown
Infrared spectrumConforms
GC>=96.0 %
StabilizerCopper
Refractive index1.5990 to 1.6030 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.