Thermo Scientific Chemicals

2-Ethylhexyl bromide, 95%, Thermo Scientific Chemicals

Catalog number: 305331000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Ethylhexyl bromide, 95%, Thermo Scientific Chemicals

Catalog number: 305331000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305331000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18908-66-2
IUPAC Name
3-(bromomethyl)heptane
Molecular Formula
C8H17Br
InChI Key
NZWIYPLSXWYKLH-UHFFFAOYNA-N
SMILES
CCCCC(CC)CBr
Appearance (Form)
Liquid
Refractive index
1.4530 to 1.4560 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
GC
>=94 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text