Dodecanedioyl dichloride, 98%, Thermo Scientific Chemicals
Dodecanedioyl dichloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dodecanedioyl dichloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 305340050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4834-98-4
IUPAC Namedodecanedioyl dichloride
Molecular FormulaC12H20Cl2O2
InChI KeyCNXXEPWXNDFGIG-UHFFFAOYSA-N
SMILESClC(=O)CCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4670 to 1.4690 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear yellow to brown
AppearanceMay darken on storage
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only