Thermo Scientific Chemicals

2,2,2-Trifluoroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 305350100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 305350100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305350100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1648-99-3
IUPAC Name
2,2,2-trifluoroethane-1-sulfonyl chloride
Molecular Formula
C2H2ClF3O2S
InChI Key
CXCHEKCRJQRVNG-UHFFFAOYSA-N
SMILES
FC(F)(F)CS(Cl)(=O)=O
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Titration Argentometric
>=94 % (ex Cl)
Infrared spectrum
Conforms
Refractive index
1.3870 to 1.3890 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text