Iron(II) acetate, 95%
Iron(II) acetate, 95%
Iron(II) acetate, 95%
Iron(II) acetate, 95%
Thermo Scientific Chemicals

Iron(II) acetate, 95%

CAS: 3094-87-9 | C4H6FeO4 | 173.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 305370100
Price (USD)
97.65
Special offer
Online exclusive
115.00
Save 17.35 (15%)
Each
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
97.65
Special offer
Online exclusive
115.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3094-87-9
IUPAC Nameλ²-iron(2+) diacetate
Molecular FormulaC4H6FeO4
InChI KeyLNOZJRCUHSPCDZ-UHFFFAOYSA-L
SMILES[Fe++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with KMnO4>=94 %
Appearance (Color)Off-white to light grey to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection