Thermo Scientific Chemicals

Iron(II) acetate, 95%, Thermo Scientific Chemicals

Catalog number: 305372500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Iron(II) acetate, 95%, Thermo Scientific Chemicals

Catalog number: 305372500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305372500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3094-87-9
IUPAC Name
λ²-iron(2+) diacetate
Molecular Formula
C4H6FeO4
InChI Key
LNOZJRCUHSPCDZ-UHFFFAOYSA-L
SMILES
[Fe++].CC([O-])=O.CC([O-])=O
Appearance (Form)
Powder
Titration with KMnO4
>=94 %
Appearance (Color)
Off-white to light grey to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text