2-Amino-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 305620050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-amino-5-bromobenzoic acid
Molecular FormulaC7H6BrNO2
InChI KeyCUKXRHLWPSBCTI-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1C(O)=O
Molecular Weight (g/mol)216.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Powder
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only