2,4-Difluorobenzylamine, 98%, Thermo Scientific Chemicals
2,4-Difluorobenzylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Difluorobenzylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 305660050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS72235-52-0
IUPAC Name(2,4-difluorophenyl)methanaminium
Molecular FormulaC7H8F2N
InChI KeyQDZZDVQGBKTLHV-UHFFFAOYSA-O
SMILES[NH3+]CC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4900 to 1.4930 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only