2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals
2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 305680050
Price (USD)/ Each
62.50
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
62.50
Add to cart
2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals
Catalog number305680050
Price (USD)/ Each
62.50
-
Add to cart
Chemical Identifiers
IUPAC Name2-cyanoethanethioamide
Molecular FormulaC3H4N2S
InChI KeyBHPYMZQTCPRLNR-UHFFFAOYSA-N
SMILESNC(=S)CC#N
Molecular Weight (g/mol)100.14
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with TBAH96.0 to 104.0 %
Appearance (Form)Powder or needles
Melting point113°C to 119°C
Appearance (Color)Beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only