Thermo Scientific Chemicals

2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: 305680250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Cyanothioacetamide, 97%, Thermo Scientific Chemicals

Catalog number: 305680250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305680250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7357-70-2
IUPAC Name
2-cyanoethanethioamide
Molecular Formula
C3H4N2S
InChI Key
BHPYMZQTCPRLNR-UHFFFAOYSA-N
SMILES
NC(=S)CC#N
Infrared spectrum
Conforms
Titration with TBAH
96.0 to 104.0 %
Appearance (Form)
Powder or needles
Melting point
113°C to 119°C
Appearance (Color)
Beige to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text