Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 305710050
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
Add to cart
Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals
Catalog number305710050
Price (USD)/ Each
105.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
124.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium methylsulfanide
Molecular FormulaCH3NaS
InChI KeyRMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES[Na+].C[S-]
Molecular Weight (g/mol)70.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granular powder
Appearance (Color)White to light beige
Titration Iodimetric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only