Thermo Scientific Chemicals

Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 305711000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium thiomethoxide, 95%, pure, Thermo Scientific Chemicals

Catalog number: 305711000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305711000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5188-07-8
IUPAC Name
sodium methylsulfanide
Molecular Formula
CH3NaS
InChI Key
RMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES
[Na+].C[S-]
Appearance (Color)
White to light beige
Appearance (Form)
Powder or granular powder
Titration Iodimetric
94 to 106 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text