Thermo Scientific Chemicals

4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 305730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Ethylbenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 305730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305730250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16331-45-6
IUPAC Name
4-ethylbenzoyl chloride
Molecular Formula
C9H9ClO
InChI Key
AVTLLLZVYYPGFX-UHFFFAOYSA-N
SMILES
CCC1=CC=C(C=C1)C(Cl)=O
Appearance (Color)
Clear colorless to yellow
GC
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.5465 to 1.5485 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text