Ethyl bromodifluoroacetate, 98%, Thermo Scientific Chemicals
Ethyl bromodifluoroacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl bromodifluoroacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 305800250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS667-27-6
IUPAC Nameethyl 2-bromo-2,2-difluoroacetate
Molecular FormulaC4H5BrF2O2
InChI KeyIRSJDVYTJUCXRV-UHFFFAOYSA-N
SMILESCCOC(=O)C(F)(F)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3860 to 1.3890 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only