Thermo Scientific Chemicals

Ethyl 6-bromohexanoate, 98%, Thermo Scientific Chemicals

Catalog number: 305921000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 6-bromohexanoate, 98%, Thermo Scientific Chemicals

Catalog number: 305921000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 305921000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25542-62-5
IUPAC Name
ethyl 6-bromohexanoate
Molecular Formula
C8H15BrO2
InChI Key
DXBULVYHTICWKT-UHFFFAOYSA-N
SMILES
CCOC(=O)CCCCCBr
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.4580 to 1.4600 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text