Methyl 4-(bromomethyl)benzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 4-(bromomethyl)benzoate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 306101000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2417-72-3
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point52°C to 57°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only