Thermo Scientific Chemicals

4-Iodobenzotrifluoride, 97%, Thermo Scientific Chemicals

Catalog number: 306180050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodobenzotrifluoride, 97%, Thermo Scientific Chemicals

Catalog number: 306180050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 306180050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
455-13-0
IUPAC Name
1-iodo-4-(trifluoromethyl)benzene
Molecular Formula
C7H4F3I
InChI Key
SKGRFPGOGCHDPC-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(I)C=C1
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5160 to 1.5200 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow to red

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text