Thermo Scientific Chemicals

Tris(4-fluorophenyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: 306340050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(4-fluorophenyl)phosphine, 98+%, Thermo Scientific Chemicals

Catalog number: 306340050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 306340050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18437-78-0
IUPAC Name
tris(4-fluorophenyl)phosphane
Molecular Formula
C18H12F3P
InChI Key
GEPJPYNDFSOARB-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Melting point
79°C to 83°C
GC
>=98.0 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text