4-(Trifluoromethoxy)benzoyl chloride, 98%, Thermo Scientific Chemicals
4-(Trifluoromethoxy)benzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethoxy)benzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 306350010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36823-88-8
IUPAC Name4-(trifluoromethoxy)benzoyl chloride
Molecular FormulaC8H4ClF3O2
InChI KeyZXKKOFJYPRJFIE-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
Appearance (Color)Clear colorless to pink
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only