Thermo Scientific Chemicals

2,6-Difluoropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 306660050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2,6-Difluoropyridine, 99%, Thermo Scientific Chemicals

Catalog number: 306660050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 306660050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1513-65-1
IUPAC Name
2,6-difluoropyridine
Molecular Formula
C5H3F2N
InChI Key
MBTGBRYMJKYYOE-UHFFFAOYSA-N
SMILES
FC1=CC=CC(F)=N1
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4360 to 1.4390 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow to pink to red-purple

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text