Methyl vinyl sulfone, 95%, stabilized, Thermo Scientific Chemicals
Methyl vinyl sulfone, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl vinyl sulfone, 95%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
50 g
Packaging:
Glass bottle
Catalog number 306730100
Price (USD)/ Each
145.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
171.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
145.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
171.00 
Save 25.35 (15%)
Add to cart
Methyl vinyl sulfone, 95%, stabilized, Thermo Scientific Chemicals
Catalog number306730100
Price (USD)/ Each
145.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
171.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS3680-02-2
IUPAC Namemethanesulfonylethene
Molecular FormulaC3H6O2S
InChI KeyWUIJTQZXUURFQU-UHFFFAOYSA-N
SMILESCS(=O)(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear colorless to yellow
Stabilizer200 ppm 4-tert-Butylphenol
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only