Thermo Scientific Chemicals

(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 307491000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

(Triphenylphosphoranylidene)acetaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 307491000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 307491000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2136-75-6
IUPAC Name
2-(triphenyl-λ⁵-phosphanylidene)acetaldehyde
Molecular Formula
C20H17OP
InChI Key
CQCAYWAIRTVXIY-UHFFFAOYSA-N
SMILES
O=CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Cream to pink to orange to brown
Titration with HClO4
96.0 to 104.0 % (on dry substance)
Water
=<4.0 % (K.F.)
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text