Thermo Scientific Chemicals

Iodoacetic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 307590010
1 g, Each
Thermo Scientific Chemicals

Iodoacetic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 307590010
1 g, Each
Quantity
Catalog number: 307590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54907-61-8
IUPAC Name
2-iodoacetyl 2-iodoacetate
Molecular Formula
C4H4I2O3
InChI Key
RBNSZWOCWHGHMR-UHFFFAOYSA-N
SMILES
ICC(=O)OC(=O)CI
Titration by coupling
96.0 to 104.0 %
Appearance (Color)
Yellow to orange
Melting point
45°C to 50°C
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text