Iodoacetic anhydride, 97%, Thermo Scientific Chemicals
Iodoacetic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodoacetic anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 307592500
Price (USD)/ Each
66.10
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
66.10
Add to cart
Iodoacetic anhydride, 97%, Thermo Scientific Chemicals
Catalog number307592500
Price (USD)/ Each
66.10
-
Add to cart
Chemical Identifiers
CAS54907-61-8
IUPAC Name2-iodoacetyl 2-iodoacetate
Molecular FormulaC4H4I2O3
InChI KeyRBNSZWOCWHGHMR-UHFFFAOYSA-N
SMILESICC(=O)OC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Titration by coupling96.0 to 104.0 %
Appearance (Color)Yellow to orange
Melting point45°C to 50°C
Appearance (Form)Powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only