Thermo Scientific Chemicals

6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 307620050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 307620050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 307620050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3222-47-7
IUPAC Name
6-methylpyridine-3-carboxylate
Molecular Formula
C7H6NO2
InChI Key
RZOKQIPOABEQAM-UHFFFAOYSA-M
SMILES
CC1=CC=C(C=N1)C([O-])=O
Appearance (Color)
White to light yellow to beige
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
209°C to 213°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text