Thermo Scientific Chemicals

cis-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals

Catalog number: 307870010
1 g, Each
Thermo Scientific Chemicals

cis-2,3-Epoxybutane, 97%, Thermo Scientific Chemicals

Catalog number: 307870010
1 g, Each
Quantity
Catalog number: 307870010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1758-33-4
Appearance (Form)
Liquid
Refractive index
1.3820 to 1.3840 (20°C, 589 nm)
GC
>=96.0 % (corrected for H2O)
Infrared spectrum
Conforms
Water
=<2.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text