2-Butynoic acid, 98%, Thermo Scientific Chemicals
2-Butynoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Butynoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 308060010
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.80 
Save 7.15 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.80 
Save 7.15 (15%)
Add to cart
2-Butynoic acid, 98%, Thermo Scientific Chemicals
Catalog number308060010
Price (USD)/ Each
41.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
48.80 
Save 7.15 (15%)
-
Add to cart
Chemical Identifiers
CAS590-93-2
IUPAC Namebut-2-ynoic acid
Molecular FormulaC4H4O2
InChI KeyLUEHNHVFDCZTGL-UHFFFAOYSA-N
SMILESCC#CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Melting point74°C to 78°C
Infrared spectrumConforms
Appearance (Form)Powder
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only