Thermo Scientific Chemicals

Perfluoro-tert-butyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 308390010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Perfluoro-tert-butyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 308390010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 308390010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2378-02-1
IUPAC Name
1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Molecular Formula
C4HF9O
InChI Key
XZNOAVNRSFURIR-UHFFFAOYSA-N
SMILES
OC(C(F)(F)F)(C(F)(F)F)C(F)(F)F
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text