Titanium(IV) fluoride, 99%, Thermo Scientific Chemicals
Titanium(IV) fluoride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(IV) fluoride, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number 308431000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-63-3
IUPAC Nametitanium(4+) tetrafluoride
Molecular FormulaF4Ti
InChI KeyXROWMBWRMNHXMF-UHFFFAOYSA-J
SMILES[F-].[F-].[F-].[F-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Titration97.5 to 102.5 % (ex F)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only