Thermo Scientific Chemicals

2,2-Dimethylsuccinic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 308760010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2-Dimethylsuccinic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 308760010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 308760010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17347-61-4
IUPAC Name
3,3-dimethyloxolane-2,5-dione
Molecular Formula
C6H8O3
InChI Key
ACJPFLIEHGFXGP-UHFFFAOYSA-N
SMILES
CC1(C)CC(=O)OC1=O
Appearance (Color)
White
Appearance (Form)
Low melting solid
Infrared spectrum
Conforms
Melting point
26°C to 30°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text