2-Ethylpyrrole, 98%, Thermo Scientific Chemicals
2-Ethylpyrrole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylpyrrole, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 308770010
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
Add to cart
2-Ethylpyrrole, 98%, Thermo Scientific Chemicals
Catalog number308770010
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1551-06-0
IUPAC Name2-ethyl-1H-pyrrole
Molecular FormulaC6H9N
InChI KeyXRPDDDRNQJNHLQ-UHFFFAOYSA-N
SMILESCCC1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4985 to 1.5025 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only