Thermo Scientific Chemicals

2-Amino-5-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 308940050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-5-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 308940050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 308940050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-08-2
IUPAC Name
2-amino-5-fluorobenzoic acid
Molecular Formula
C7H6FNO2
InChI Key
FPQMGQZTBWIHDN-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C=C1C(O)=O
Appearance (Color)
Light yellow to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
181°C to 185°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text