Appearance (Form)Powder and/or chunks or low melting solid
GC>=97.5 %
Appearance (Color)White to yellow
Melting point29°C to 37°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.