Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 309130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: 309130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 309130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
327-78-6
IUPAC Name
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene
Molecular Formula
C8H3ClF3NO
InChI Key
NBJZEUQTGLSUOB-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC=C1Cl)N=C=O
Appearance (Form)
Powder and/or chunks or low melting solid
GC
>=97.5 %
Appearance (Color)
White to yellow
Melting point
29°C to 37°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text