Thermo Scientific Chemicals

Tetracontane, 98%, Thermo Scientific Chemicals

Catalog number: 309160025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetracontane, 98%, Thermo Scientific Chemicals

Catalog number: 309160025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 309160025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4181-95-7
IUPAC Name
tetracontane
Molecular Formula
C40H82
InChI Key
KUPLEGDPSCCPJI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Melting point
80°C to 83°C
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text