Tetracontane, 98%, Thermo Scientific Chemicals
Tetracontane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetracontane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 309165000
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.00 
Save 14.35 (15%)
-
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.00 
Save 14.35 (15%)
Add to cart
Tetracontane, 98%, Thermo Scientific Chemicals
Catalog number309165000
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
95.00 
Save 14.35 (15%)
-
Add to cart
Chemical Identifiers
CAS4181-95-7
IUPAC Nametetracontane
Molecular FormulaC40H82
InChI KeyKUPLEGDPSCCPJI-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point80°C to 83°C
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only